Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD98 Mouse anti-Human, BB515, Clone: UM7F8, BD

Catalog No. BDB565103 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565103 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565103 Supplier BD Biosciences Supplier No. 565103

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

CD98 is a disulfide-linked heterodimer composed of a ∽80-85 kDa, type II membrane glycoprotein heavy chain, (CD98hc), and ∽40-45 kDa light chain. The UM7F8 monoclonal antibody specifically recognizes CD98hc, which is also known as 4F2 heavy chain antigen (4F2hc). CD98 is broadly expressed on hematopoietic cells, including peripheral blood lymphocytes, monocytes and granulocytes (low), as well as non-hematopoietic cells, eg, intestinal epithelial cells. CD98 expression is upregulated on activated and proliferating cells. CD98hc is encoded by the SLC3A2 [solute carrier family 3 (amino acid transporter heavy chain), member 2] gene. CD98 reportedly functions in transmembrane amino acid transport and in the regulation of integrin signaling which are involved in the regulation of cellular activation, proliferation, and survival. The UM7F8 antibody is reportedly a functional antibody that can costimulate T cell proliferative responses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD98
Applications Flow Cytometry
Classification Monoclonal
Clone UM7F8
Conjugate Brilliant Blue 515 (BB515)
Description SLC3A2; CD98HC; 4F2; 4F2hc; MDU1; NACAE; 4T2HC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide
Host Species Mouse
Immunogen Human T-leukemic and Thymic Cell Lines
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.