Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-CD99 Clone: O662,

Catalog No. MABF927MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF927MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF927MI Supplier MilliporeSigma™ Supplier No. MABF927
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects CD99 Clone: O662 in Human samples, and it is validated for Activity Assay, Flow Cytometry, Western Blotting

CD99 antigen (UniProt P14209; also known as 12E7, E2 antigen, HBA-71 antigen, Protein MIC2, T-cell surface glycoprotein E2, Surface antigen MIC2) is encoded by the CD99 (also known as HBA71, MIC2, MIC2X, MIC2Y, MSK5X) gene (Gene ID 4267) in human. Originally identified as a thymus-leukemia-associated antigen, CD99 is a transmembrane glycoprotein expressed on many hematopoietic and non-hematopoietic cell types. CD99 is involved in cellular adhesion and migration as well as in survival. Antibody-mediated cross-linking of cell surface CD99 is shown to induce apoptotic signals in Jurkat T cells and essential thrombocythemia (ET) cell lines in vitro, and several CD99 monoclonal antibodies show in vivo efficacy in ET mouse models. CD99 is initially produced with a signal peptide sequence (a.a. 1-22), the removal of which yields the mature CD99 with a large extracellular domain (a.a. 23-122), followed by a transmembrane segment (a.a. 123-147) and a short cytoplasmic tail (a.a. 148-185).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD99
Applications Functional Assay, Flow Cytometry, Western Blot
Classification Monoclonal
Clone O662
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG3κ antibody in PBS without preservatives.
Gene Accession No. P14209
Gene Symbols CD99, HBA71, MIC2, MIC2X, MIC2Y, MSK5X
Host Species Mouse
Immunogen Human thymocytes.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation, Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001116370
Test Specificity Clone 0662 detects both the 32kDa glycosylated (plasma membrane) and the 28kDa non-glycosylated (intracellular) forms of human CD99/E2 (Gelin, C., et al. (1989). EMBO J. 8(11):3253-3259). Note that the clone was originally designated as 0662 (zero six six two) by Bernard, A., et al. (1988), but subsequently cited in some publications as O662 with a captial letter O.
Target Species Human
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Isotype IgG3 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.