Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD99 Mouse anti-Human, PE, Clone: TÜ12, BD

Catalog No. BDB555689 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555689 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555689 Supplier BD Biosciences Supplier No. 555689
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recognizes CD99, also referred to as E2 antigen, a 32kDa sialoglycoprotein expressed on all leukocyte lineages. The E2 antigen is the MIC2 gene product and is differentially expressed during T- and B-lymphoid and granulocytic development, with higher densities being expressed during early hematopoietic stages. Mature granulocytes express very little or no CD99. E2 has been shown to be involved in T-cell adhesion processes and is suggested to have a functional role in hematopoietic adhesion pathways. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: TU12
Isotype: IgG2a κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD99
Applications Flow Cytometry
Classification Monoclonal
Clone TÜ12
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.