Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdc2 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB324125UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB324125UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB324125UL Supplier MilliporeSigma Supplier No. AB324125UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-Cdc2 Antibody, phospho-specific (Tyr15) (Rabbit Polyclonal Antibody) validated in western blotting to detect Cdc2 also known as Cell Division Cycle 2, p34 Protein Kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin in 10mM Hepes, pH 7.5, with 150mM NaCl, 100μg/mL BSA and 50% glycerol.
Gene Symbols CDC2; CDK1; DKFZp686L20222; MGC111195; CDC28A; EC 2.7.11.22; EC 2.7.11.23
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids surrounding the phosphoTry15 of human, rat, mouse and Xenopus CDC2.
Purification Method Affinity Purified
Quantity 25 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001786.2; NM_033379.2
Target Species Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.