Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CDC25C Ab-7, Clone: 25C07 (TC-113), Epredia™ LabVision

Catalog No. MS1187PCS Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
100 μL
200 μL
5 Slides
500 μL
7 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS1187PCS 5 Slides Unconjugated
MS1187B0 100 μL Biotin 200μg/mL
MS1187B 1 mL Biotin 200μg/mL
MS1187P1 500 μL Unconjugated 200μg/mL
MS1187P1ABX 100 μL Unconjugated 1mg/mL
MS1187PABX 200 μL Unconjugated 1mg/mL
MS1187R7 7 mL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 missing translation for 'options'
Catalog No. MS1187PCS missing translation for 'mfr' Epredia missing translation for 'supplierNo' MS1187PCS

Mouse Monoclonal Antibody

Cyclin-dependent kinases can be activated by CDC25, which removes inhibitory phosphates from tyrosine and threonine residues. At least three CDC25 genes (CDC25A, CDC25B, and CDC25C) have been identified in humans. Whereas CDC25A and CDC25B are expressed throughout the cell cycle, with peak expression in G1 for CDC25A and in both G1-S-phase and G2 for CDC25B, CDC25C is predominantly expressed in G2.

Host Species: Mouse
Clone: 25C07 (TC-113)
Isotype: IgG1/κ
Cross Reactivity: Human. Does not react with mouse and rat. Others-not tested.
Epitope: aa222-246
Immunogen: Recombinant human CDC25C protein.
Molecular Weight: 53-56kDa
Positive Control: HeLa, HT29, or LS174T cells. Breast CA.
Cellular Localization: Cytoplasmic

Ab-7 is excellent for the staining of formalin-fixed, paraffin-embedded tissues.

Immunofluorescence, Immunohistochemistry (Formalin/Paraffin), Immunoprecipitation, Western Blotting

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Antigen CDC25C Ab-7
Classification Monoclonal
Clone 25C07 (TC-113)
Concentration
Conjugate Unconjugated
Host Species Mouse
Quantity 5 Slides
Primary or Secondary Primary
Target Species Human
Isotype IgG1 κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.