Learn More
CDC42 Mouse, Unlabeled, Clone: 44, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610929
Description
Rho family members are small GTP binding proteins that serve as molecular switches for a number of biological processes. They cycle between active GTP-bound and inactive GDP-bound states. CDC42 is a Rho family protein that was identified in membranes of human platelets and placenta. It is the homologue of CDC42Sc, which regulates initiation of bud-site assembly in Saccharomyces cerevisiae. CDC42 regulates the function of the mammalian actin cytoskeleton, allowing for efficient cytokinesis and cell morphogenesis. CDC42 and Rac1, a Ras-related GTPase, activate MEKK1, a JNK kinase kinase, which leads to the activation of several downstream components of the MAP kinase cascade leading to activation of PAK65, a CDC42- and Rac-binding protein. PAK65 interacts with CDC42/Rac1, mediates their interaction with MEKK1, and enhances MEKK1 catalytic activity. Chronic activation of CDC42 has been shown to induce malignant cellular transformation. Due to sequence homology among various small GTPase family members, potential cross-reactivity could be observed with this antibody. BD Pharmingen has not performed such cross-reactivity testing.
Immunofluorescence, Western Blotting

Specifications
CDC42 | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human CDC42 aa. 1-191 | |
150 μg | |
Cell Biology | |
Canine, Human, Mouse, Rat | |
IgG1 |
Western Blot | |
44 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.