Learn More
CDC42 effector protein (Rho GTPase binding) 1 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant CDC42EP1.
Supplier: Abnova Corporation H00011135A01
Description
CDC42 is a member of the Rho GTPase family that regulates multiple cellular activities, including actin polymerization. The protein encoded by this gene is a CDC42 binding protein that mediates actin cytoskeleton reorganization at the plasma membrane. This protein is secreted and is primarily found in bone marrow. [provided by RefSeq
Sequence: MPGPQGGRGAATMSLGKLSPVGWVSSSQGKRRLTADMISHPLGDFRHTMHVGRGGDVFGDTSFLSNHGGSSGSTHRSPRSFLAKKLQLVRRVGAPPRRMASPPAPSPAPPAISPIIKNAISLPQLNQAAYDSLVVGKLSFDSSPTSSTDGHSSYGLDSGFCTISRLPRSEKPHDRDRDGSFPSEPGLRRSDSLLSFRLDLDLGPSLLSELLGVMSLPEAPAAETPAPAANPPAPTANPTGPAANPPATTANPPAPAATPTGPAANPPAPAASSTPHGHCPNGVTAGLGPVAEVKSSPVGGGPRGPAGPALGRHWGAGWDGGHHYPEMDARQERVEVLPQARASWESLDEEWRAPQAGSRTPVPSTVQANTFEFADAEEDDEVKVSpecifications
CDC42 effector protein (Rho GTPase binding) 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length recombinant CDC42EP1. | |
CDC42EP1 | |
BORG5/CEP1/MGC15316/MSE55 | |
Mouse | |
50 μL | |
Yes | |
11135 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
BC009356 | |
CDC42EP1 | |
CDC42EP1 (AAH09356, 1 a.a. ∼ 384 a.a) full-length recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.