Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC5L, Mouse, Clone: 3C12, Abnova™

Catalog No. 89021015 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-015 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-015 Supplier Abnova Corporation Supplier No. H00000988M08
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CDC5L.

The protein encoded by this gene shares a significant similarity with Schizosaccharomyces pombe cdc5 gene product, which is a cell cycle regulator important for G2/M transition. This protein has been demonstrated to act as a positive regulator of cell cycle G2/M progression. It was also found to be an essential component of a non-snRNA spliceosome, which contains at least five additional protein factors and is required for the second catalytic step of pre-mRNA splicing. [provided by RefSeq

Sequence: ILLGGYQSRAMGLMKQLNDLWDQIEQAHLELRTFEELKKHEDSAIPRRLECLKEDVQRQQEREKELQHRYADLLLEKETLKSKF

Specifications

Antigen CDC5L
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3C12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CDC5L.
Formulation PBS with no preservative; pH 7.4
Gene CDC5L
Gene Accession No. NM_001253
Gene Alias CEF1/KIAA0432/PCDC5RP/dJ319D22.1/hCDC5
Gene Symbols CDC5L
Host Species Mouse
Immunogen CDC5L (NP_001244, 719 a.a. ∼ 802 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 988
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.