Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cell division cycle 6 homolog (S. cerevisiae), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017499 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-499 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-499 Supplier Abnova Corporation Supplier No. H00000990D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human CDC6 protein.

The protein encoded by this gene is highly similar to Saccharomyces cerevisiae Cdc6, a protein essential for the initiation of DNA replication. This protein functions as a regulator at the early steps of DNA replication. It localizes in cell nucleus during cell cyle G1, but translocates to the cytoplasm at the start of S phase. The subcellular translocation of this protein during cell cyle is regulated through its phosphorylation by Cdks. Transcription of this protein was reported to be regulated in response to mitogenic signals through transcriptional control mechanism involving E2F proteins. [provided by RefSeq

Sequence: MPQTRSQAQATISFPKRKLSRALNKAKNSSDAKLEPTNVQTVTCSPRVKALPLSPRKRLGDDNLCNTPHLPPCSPPKQGKKENGPPHSHTLKGRRLVFDNQLTIKSPSKRELAKVHQNKILSSVRKSQEITTNSEQRCPLKKESACVRLFKQEGTCYQQAKLVLNTAVPDRLPAREREMDVIRNFLREHICGKKAGSLYLSGAPGTGKTACLSRILQDLKKELKGFKTIMLNCMSLRTAQAVFPAIAQEICQEEVSRPAGKDMMRKLEKHMTAEKGPMIVLVLDEMDQLDSKGQDVLYTLFEWPWLSNSHLVLIGIANTLDLTDRILPRLQAREKCKPQLLNFPPYTRNQIVTILQDRLNQVSRDQVLDNAAVQFCARKVSAVSGDVRKALDVCRRAIEIVESDVKSQTILKPLSECKSPSEPLIPKRVGLIHISQVISEIDGNRMTLSQEGAQDSFPLQQKILVCSLMLLIRQLKIKEVTLGKLYEAYSKVCRKQQVAAVDQSECLSLSGLLEARGILGLKRNKETRLTKVFFKIEEKEIEHALKDKALIGNILATGLP

Specifications

Antigen cell division cycle 6 homolog (S. cerevisiae)
Applications Immunofluorescence, In situ PLA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human CDC6 protein.
Formulation PBS with no preservative; pH 7.4
Gene CDC6
Gene Accession No. BC025232
Gene Alias CDC18L/HsCDC18/HsCDC6
Gene Symbols CDC6
Host Species Rabbit
Immunogen CDC6 (AAH25232.1, 1 a.a. ∼ 560 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 990
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.