Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDCA7L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA523443
Description
CDCA7L Polyclonal Antibody for Western Blot
Cell division cycle-associated 7-like protein (or RAM2) is a transcription factor which plays a role in transcriptional regulation and apoptotic signaling pathways. It acts as a repressor that inhibits monoamine oxidase A (MAOA) activity and gene expression by binding to promoter. It also plays an important oncogenic role in mediating the full transforming effect of MYC in medulloblastoma cells. It may act downstream of P38-kinase and BCL-2, but upstream of CASP3/caspase-3 as well as CCND1/cyclin D1 and E2F1.
Specifications
CDCA7L | |
Polyclonal | |
Unconjugated | |
CDCA7L | |
BC006933; CDCA7L; cell division cycle associated 7 like; cell division cycle associated 7-like; cell division cycle-associated 7-like protein; HR1; JPO2; Protein JPO2; R1; RAM2; transcription factor RAM2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
217946, 55536 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
Q922M5, Q96GN5 | |
CDCA7L | |
A portion of amino acid 170 to 220 of human CDCA7L/RAM2. | |
100 μg | |
Primary | |
Human, Mouse, Monkey | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction