Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Cdh1 Clone: DH01,

Catalog No. CC43100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
CC43100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC43100UG Supplier MilliporeSigma™ Supplier No. CC43100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Cdh1 Clone: DH01 in Human samples, and it is validated for Immunoprecipitation

Cdh1, a homolog of the yeast Cdc20, is a ∽55kDa protein required for the degradation of anaphase-promoting complex (APC)-dependent proteolysis. Cdh1 acts as a limiting, substrate-specific activator of APC-dependent proteolysis and it is required for the degradation of APC substrates such as Clb2 and Ase1. Cdh1 plays an important role in mitotic regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdh1
Applications Immunoprecipitation
Classification Monoclonal
Clone DH01
Conjugate Unconjugated
Formulation In 10mM PBS, 0.2% BSA, pH 7.4.
Host Species Mouse
Immunogen recombinant human Cdh1 protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽55kDa Cdh-1 protein in Rat-1 cells.
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.