Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDH17, Mouse, Clone: 3H2, Abnova™

Catalog No. 89021019 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-019 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-019 Supplier Abnova Corporation Supplier No. H00001015M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CDH17.

This gene is a member of the cadherin superfamily, genes encoding calcium-dependent, membrane-associated glycoproteins. The encoded protein is cadherin-like, consisting of an extracellular region, containing 7 cadherin domains, and a transmembrane region but lacking the conserved cytoplasmic domain. The protein is a component of the gastrointestinal tract and pancreatic ducts, acting as an intestinal proton-dependent peptide transporter in the first step in oral absorption of many medically important peptide-based drugs. The protein may also play a role in the morphological organization of liver and intestine. Alternative splicing results in multiple transcript variants. [provided by RefSeq

Sequence: EGKFSGPLKPMTFSIYEGQEPSQIIFQFKANPPAVTFELTGETDNIFVIEREGLLYYNRALDRETRSTHNLQVAALDANGIIVEGPVPITIEVKDINDNRPTFLQSKY

Specifications

Antigen CDH17
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 3H2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CDH17.
Formulation PBS with no preservative; pH 7.4
Gene CDH17
Gene Accession No. NM_004063
Gene Alias CDH16/FLJ26931/HPT-1/HPT1/MGC138218/MGC142024
Gene Symbols CDH17
Host Species Mouse
Immunogen CDH17 (NP_004054, 24 a.a. ∼ 131 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1015
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.