Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDH6, Mouse, Clone: 2F2, Abnova™

Catalog No. 89021018 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-018 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-018 Supplier Abnova Corporation Supplier No. H00001004M05
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CDH6.

This gene encodes a type II classical cadherin from the cadherin superfamily. The encoded membrane protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Cadherins mediate cell-cell binding in a homophilic manner, contributing to the sorting of heterogeneous cell types and the maintenance of orderly structures such as epithelium. Strong transcriptional expression of this gene has been observed in hepatocellular and renal carcinoma cell lines, suggesting a possible role in metastasis and invasion. [provided by RefSeq

Sequence: DKDDPYSGHQFSFSLAPEAASGSNFTIQDNKDNTAGILTRKNGYNRHEMSTYLLPVVISDNDYPVQSSTGTVTVRVCACDHHGNMQSCHAEALIHPTGLS

Specifications

Antigen CDH6
Applications ELISA, KnockDown, Western Blot
Classification Monoclonal
Clone 2F2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CDH6.
Formulation PBS with no preservative; pH 7.4
Gene CDH6
Gene Accession No. NM_004932
Gene Alias KCAD
Gene Symbols CDH6
Host Species Mouse
Immunogen CDH6 (NP_004923, 513 a.a. ∼ 612 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1004
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.