Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDIPT Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526054
Description
This antibody is predicted to react with rat based on sequence homology.
CDIPT is a protein which is phosphatidylinositol breakdown products that are ubiquitous second messengers that function downstream of many G protein-coupled receptors and tyrosine kinases regulating cell growth, calcium metabolism, and protein kinase C activity. Two enzymes, CDP-diacylglycerol synthase and phosphatidylinositol synthase, are involved in the biosynthesis of phosphatidylinositol. Phosphatidylinositol synthase, a member of the CDP-alcohol phosphatidyl transferase class-I family, is an integral membrane protein found on the cytoplasmic side of the endoplasmic reticulum and the Golgi apparatus.
Specifications
CDIPT | |
Polyclonal | |
Unconjugated | |
O14735, Q8VDP6 | |
CDIPT | |
KLH conjugated synthetic peptide between 99-125 amino acids from the central region of human CDIPT | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
CDIPT | |
PIS, PIS1, PI synthase, PtdIns synthase, phosphatidylinositol synthase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10423, 52858 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction