Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk1/Cdc2 (PSTAIR) Rabbit anti-Amphibian, Chicken, Human, Mouse, Rat, Sheep, Drosophila, Monkey, Canine, Polyclonal, MilliporeSigma™

Catalog No. 06923MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-923-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-923-MI Supplier MilliporeSigma Supplier No. 06923
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Cdk1/Cdc2 (PSTAIR) Antibody is a high quality Rabbit Polyclonal Antibody for the detection of Cdk1/Cdc2 (PSTAIR) and has been validated in immunocytochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdk1/Cdc2 (PSTAIR)
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit serum in Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols CDC2; CDK1; MGC111195; DKFZp686L20222; CDC28A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues 42-57 of human cdc2 kinase. The immunogen is identical in a wide range of species.
Purification Method Antigen affinity purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001786
Target Species Amphibian, Canine, Chicken, Drosophila, Human, Monkey, Mouse, Rat, Sheep
Content And Storage 2-8°C for 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.