Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk1 Mouse, Unlabeled, Clone: 1, BD

Catalog No. BDB610037 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610037 50 μg
BDB610038 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610037 Supplier BD Biosciences Supplier No. 610037
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cdk1, also known as p34 [cdc2], is a ubiquitously expressed serine/threonine protein kinase. Cdk1/cdc2 has been identified as the catalytic subunit of the maturation-promoting factor (MPF), while cyclin B acts as the regulatory subunit. The binding of these two subunits is critical to the transition into M-phase of the mammalian cell cycle, and this factor's role is regulated by a series of phosphorylations and dephosphorylations. After binding to cyclin B, cdk1/cdc2 is phosphorylated on Thr-14, by Myt1, and Tyr-15, by wee1 or mik1, yielding an inactive pre-MPF complex. Phosphorylation of cdk1/cdc2 on Thr-161 is performed by a cdk7/cyclin H complex and is necessary for activation of the cdc2 complex. Dephosphorylation of Thr-14 and Tyr-15 by CDC25 occurs at the prophase/metaphase transition and completes activation of the cdc2/cyclin B complex, initiating the cell's entry into mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdk1
Applications Western Blot
Classification Monoclonal
Clone 1
Concentration 250μg/mL
Conjugate Unconjugated
Description Cdc2, p34
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human p34 [cdc2] aa. 283 to 297
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.