Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk1 Mouse anti-Chicken, Human, Mouse, Rat, Xenopus, Clone: A17.1.1, MilliporeSigma™

Catalog No. MAB8878MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8878MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8878MI Supplier MilliporeSigma Supplier No. MAB8878
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Cdk1 Antibody, clone A17.1.1 is a Mouse Monoclonal Antibody for detection of Cdk1 also known as p34cdc2, CDC2 and has been validated in immunofluoresence, immunoprecipitation, EA, western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdk1
Applications Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone A17.1.1
Conjugate Unconjugated
Formulation Purified immunoglobulin. 0.2mg/mL in 10mM PBS, pH 7.4, containing 0.2% BSA and 0.09% Sodium Azide.
Gene Accession No. NM_001786.2; NM_033379.2
Gene Symbols CDC2, CDK1, DKFZp686L20222, MGC111195, CDC28A, EC 2.7.11.22, EC 2.7.11.23
Host Species Mouse
Immunogen Raised against Xenopus p34cdc2.
Quantity 50 μg
Primary or Secondary Primary
Target Species Chicken, Human, Mouse, Rat, Xenopus species
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.