Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk2 Mouse, Unlabeled, Clone: 55, BD

Catalog No. BDB610146 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610146 150 μg
BDB610145 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610146 Supplier BD Biosciences Supplier No. 610146

Mouse Monoclonal Antibody

Cyclin-dependent kinase 2 (Cdk2) is a member of a family of cdc2-related cell cycle protein kinases. Cdk2 shares 60% identity with cdc2 and its activity is regulated by phosphorylation in a similar fashion. Cdk2 is expressed earlier in the cell cycle than is cdc2. Like p34 [cdc2], p33 [cdk2] associates with Cyclin A in human cells. However, kinase activity associated with Cyclin A-Cdk2 is present in S phase, whereas, the kinase activity associated with Cyclin A-cdc2 is found only in G2. Cdk2 can also complex with cyclins E, D1, and D3. It is not known if the D cyclins can form active complexes with Cdk2. Cyclin E-Cdk2 kinase is active in the G1 and S phases of the cell cycle and is important (as is Cyclin A-Cdk2) for the progression from G1 to S phase. The levels of Cyclin A-Cdk2 are maximal at the G1/S transition and both Cdk2 and Cyclin A associate with DNA in the initiation complex during replication. The Rb protein has been identified as a substrate for Cdk2-Cyclin E and/or Cdk2-Cyclin A in vivo. This observation is supported by further evidence which shows that Cdk2 is activated and specifically localized to the nucleus during late G1, S phase, and G2.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdk2
Applications Western Blot
Classification Monoclonal
Clone 55
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.