Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk2 Mouse anti-Human, Mouse, Xenopus, Clone: AN4.3, MilliporeSigma™

Catalog No. 05596MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-596-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-596-MI Supplier MilliporeSigma Supplier No. 05596
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Cdk2 with Anti-Cdk2 Antibody, clone AN4.3 (Mouse Monoclonal Antibody), that has been shown to work in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdk2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone AN4.3
Conjugate Unconjugated
Formulation 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide, and 30% glycerol.
Gene Accession No. NM_001798
Gene Symbols CDK2, p33(CDK2)
Host Species Mouse
Immunogen Recombinant human cdk2
Purification Method Protein G chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Mouse, Xenopus species
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.