Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk4 Mouse, Unlabeled, Clone: 97, BD

Catalog No. BDB610147 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610147 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610147 Supplier BD Biosciences Supplier No. 610147

Mouse Monoclonal Antibody

The Cdk4 kinase is a cell cycle-regulated cyclin-dependent kinase that associates with cyclins D1, D2, and D3. In macrophages and fibroblasts, Cdk4-Cyclin D1 complexes are predominant. Much like Cdk2 and Cdc2, the activity of Cdk4 is regulated by association with its respective kinase and by phosphorylation of specific threonine residues. The cdk-activating kinase (CAK) is a regulator of cdk-cyclin activity by virtue of its ability to phosphorylate single threonine residues on Cdk2, Cdc2, and Cdk4. Active Cdk4-Cyclin D1 complexes are capable of phosphorylating the retinoblastoma gene product (pRb), but not histone H1 or casein. Formation of the Cdk4-Cyclin D holoenzyme and phosphorylation of the catalytic subunit appear to be independently regulated. Therefore, other cellular factors may be necessary for stabilization of this protein complex in order to facilitate the phosphorylation of Cdk4. An inhibitor of Cdk4 kinase activity, known as p16, has been identified and partially characterized. The p16 gene appears to be altered in many tumors, suggesting that this is a tumor suppressor gene. It is thought that p16 inhibition of Cdk4-Cyclin D interferes with cell cycle progression by preventing phosphorylation of pRb by Cdk4 and the subsequent release of factors that activate transcription.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdk4
Applications Western Blot
Classification Monoclonal
Clone 97
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.