Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk4 (N-Term) Mouse anti-Human, Unlabeled, Clone: DCS-35, BD

Catalog No. BDB559693 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559693 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559693 Supplier BD Biosciences Supplier No. 559693

Mouse Monoclonal Antibody

Cyclins, cyclin-dependent kinases (Cdks), and cyclin-dependent kinase inhibitors (CdkIs) are essential for cell-cycle control in eukaryotes (reviewed in 1). Cyclins, regulatory subunits, bind to cyclin-dependent kinases (Cdks), catalytic subunits, to form active cyclin-Cdk complexes. Cdk subunits by themselves are inactive and binding to a cyclin is required for their activity. Cyclins A, B1, D and E undergo periodic synthesis and degradation, thereby providing a mechanism to regulate Cdk activity throughout the cell cycle. Additionally, Cdk activity is further regulated by activating and inhibitory phosphorylations, and small proteins (p15, p16, p18, p19, p21 and p27), called CdkIs, that bind to cyclins, Cdks, or cyclin-Cdk complexes. Cdk4 was originally called PSK-J3,2 and following demonstration of its association with D-type cyclins, became known as Cdk4.2 D-type cyclins also associate with Cdks 2 and 5, although Cdk4 appears to be the most abundant partner. The D-type cyclins (D1, D2, and D3) are expressed in response to growth factors or mitogens, and rapidly degrade when mitogens are withdrawn. D cyclins appear to promote G0 to G1 transitions and the rate of G1 progression. For example, cyclin D-Cdk4 and cyclin D-Cdk6
complexes phosphorylate the retinoblastoma protein (Rb) which removes the G1 phase block caused by underphosphorylated Rb. Cdk4 has a molecular weight of ∽32kDa.
This antibody is routinely tested by western blot and bioimaging analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdk4 (N-Term)
Applications Western Blot
Classification Monoclonal
Clone DCS-35
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Cdk4
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.