Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk5 Mouse anti-Drosophila, Human, Mouse, Rat, Clone: DC17, MilliporeSigma™

Catalog No. 05364MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-364-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-364-MI Supplier MilliporeSigma Supplier No. 05364
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Cdk5 Antibody, clone DC17 is a high quality Mouse Monoclonal Antibody for the detection of Cdk5 and has been validated in immunoprecipitation, western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdk5
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone DC17
Conjugate Unconjugated
Formulation 0.1M Tris-glycine pH 7.4, 0.15M NaCl, containing 0.05% sodium azide
Gene Accession No. NP_004926
Gene Symbols CDK5, PSSALRE
Host Species Mouse
Immunogen Recombinant human cdk5
Purification Method Protein G chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Drosophila, Human, Mouse, Rat
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.