Learn More
CDK5R2, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human CDK5R2 peptide using ARM Technology.
Supplier: Abnova Corporation H00008941K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The protein encoded by this gene is a neuron-specific activator of CDK5 kinase. It associates with CDK5 to form an active kinase. This protein and neuron-specific CDK5 activator CDK5R1/p39NCK5A both share limited similarity to cyclins, and thus may define a distinct family of cyclin-dependent kinase activating proteins. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
CDK5R2 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
CDK5R2 | |
CDK5R2 | |
A synthetic peptide of human CDK5R2 | |
RUO | |
8941 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
cyclin-dependent kinase 5, regulatory subunit 2 (p39) | |
CDK5R2 | |
NCK5AI, P39, p39nck5ai | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.