Learn More
Description
CDK7 is cdk activating kinase associated with the 34kDa cyclin H protein, and a 36kDa unnamed protein. CDK7/cyclin H is an integral part of the general transcription factor complex TFIIH; hence is important for activated transcription of polymerase II-dependent promoters and DNA repair. CDK7 is nuclear at all stages of the cell division cycle, except for mitosis indicating that its activity is required for key CDKs operating in distinct cell cycle phases.
Host Species: Mouse
Clone: MO-1.1
Isotype: IgG2b
Cross Reactivity: Human and Rat. Does not react with mouse. Others-not known.
Epitope: 221 amino acid fragment of C-terminus
Immunogen: A recombinant 221 amino acid fragment of the C-terminus human p40MO15 protein.
Molecular Weight: 40kDa
Positive Control: HT29 and LS174T cells
Cellular Localization: Nuclear and cytoplasmic
Ab-1 is well suited for multiple biochemical applications including immunoprecipitation of kinase activity
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Kinase Assay, Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
| Antigen | Cdk7/ p40MO15/CAK Ab-1 |
| Concentration | 200μg/mL |
| Host Species | Mouse |
| Immunogen | A recombinant 221 amino acid fragment of the C-terminus human p40MO15 protein. |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Target Species | Human, Rat |
| Isotype | IgG2b |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.