Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDKL1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA514455
Description
This gene product is a member of a large family of CDC2-related serine/threonine protein kinases. It accumulates primarily in the nucleus. Alternative splice variants encoding different protein isoforms have been described, but their full-length nature has not been determined.
Specifications
CDKL1 | |
Polyclonal | |
Unconjugated | |
Q00532, Q8CEQ0 | |
CDKL1 | |
KLH conjugated synthetic peptide between 285-316 amino acids from the C-terminal region of human CDKL1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
CDKL1 | |
Serine/threonine-protein kinase KKIALRE, Cyclin-dependent kinase-like 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
71091, 8814 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction