Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
cyclin-dependent kinase-like 4, Mouse, Clone: 6G8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CDKL4.
Supplier: Abnova Corporation H00344387M03
Description
Sequence: TGQPLWPGKSDVDQLYLIIRTLGKLIPRHQSIFKSNGFFHGISIPEPEDMETLEEKFSDVHPVALNFMKGCLKMNPDDRLTCSQLLESSYFDSFQEAQIKSpecifications
cyclin-dependent kinase-like 4 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001009565 | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
344387 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
6G8 | |
Mouse monoclonal antibody raised against a partial recombinant CDKL4. | |
CDKL4 | |
CDKL4 | |
CDKL4 (AAV63975, 196 a.a. ∼ 295 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Kinases | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction