Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDS1, Mouse, Clone: 2D10, Abnova™

Catalog No. 89021038 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-038 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-038 Supplier Abnova Corporation Supplier No. H00001040M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CDS1.

Breakdown products of phosphoinositides are ubiquitous second messengers that function downstream of many G protein-coupled receptors and tyrosine kinases regulating cell growth, calcium metabolism, and protein kinase C activity. This gene encodes an enzyme which regulates the amount of phosphatidylinositol available for signaling by catalyzing the conversion of phosphatidic acid to CDP-diacylglycerol. This enzyme is an integral membrane protein localized to two subcellular domains, the matrix side of the inner mitochondrial membrane where it is thought to be involved in the synthesis of phosphatidylglycerol and cardiolipin and the cytoplasmic side of the endoplasmic reticulum where it functions in phosphatidylinositol biosynthesis. Two genes encoding this enzyme have been identified in humans, one mapping to human chromosome 4q21 and a second to 20p13. [provided by RefSeq

Sequence: MLELRHRGSCPGPREAVSPPHREGEAAGGDHETESTSDKETDIDDRYGDLDSRTDSDIPEIPPSSDRTPEILKKALSGLSSRWKNWWIRGILTLTMIS

Specifications

Antigen CDS1
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2D10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CDS1.
Formulation PBS with no preservative; pH 7.4
Gene CDS1
Gene Accession No. NM_001263
Gene Alias CDS
Gene Symbols CDS1
Host Species Mouse
Immunogen CDS1 (NP_001254, 1 a.a. ∼ 98 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1040
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.