Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDw210 Rat anti-Human, PE, Clone: 3F9, BD

Catalog No. BDB556013 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556013 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556013 Supplier BD Biosciences Supplier No. 556013
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Reaacts with 90-110kDa protein expressed on T cells, B cells, NK cells, and monocyte/macrophages. It belongs to the class II cytokine receptor family which includes IFNγR (CDw119), IFNα/βR (CD118) and tissue factor (CD142). IL-10R has been found to play a role in signal transduction by activating specific STAT proteins (STAT3 and STAT1) and inducing gene transcription. Other reports indicate that IL-10 protein binding to NK cells appears to be stimulatory, in contrast to its binding to T cells and monocytes/macrophages. Clone 3F9 is specific for human IL-10R and its binding to the receptor can be blocked by recombinant human IL-10 (rhIL-10). This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: 3F9
Isotype: IgG2a κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CDw210
Applications Flow Cytometry
Classification Monoclonal
Clone 3F9
Conjugate PE
Description IL-10 receptor
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Human IL-10R alpha Recombinant Protein
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.