Learn More
CDw329 Mouse anti-Human, FITC, Clone: E10-286, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 550906
Description
Reacts with Siglec-9, formerly known as Siglec-Y, expressed on peripheral blood monocytes, granulocytes and NK cells. Siglecs (sialic acid/immunolgobulin/lectin) are a family of I-type lectins binding to sialic acids on the cell surface. They are a family
Host Species: Mouse
Clone: E10-286
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry

Specifications
CDw329 | |
Monoclonal | |
FITC | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
E10-286 | |
Siglec-9; Siglec Y; FOAP-9; OBBP-LIKE | |
Mouse | |
100 Tests | |
Cell Biology | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.