Learn More
CDw93 (C1qRp) Mouse anti-Human, Unlabeled, Clone: R3, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 551454
Description
The immunogen used to raise R3 was C1q-binding protein preparation as described. Human C1qRp is a 631 a.a. protein (∽66.5kD) protein that is highly expressed on monocytes/macrophages, neutrophil granulocytes but not on T and B lymphocytes. C1qRp binds C1q, the recognition subunit of the first component (C1) of the complement pathway, as well as MBL (Mannose-binding-lectin) and SPA (Pulmonary Surfactant Protein A). Human C1qRp is involved in the C1q-mediated enhancement of phagocytosis. R3 is suitable to detect C1qRp
expression on cells of myeloid lineage by flow cytometry, C1qRp in cellular lysates by Western blotting or immunoprecipitation. Pretreatment of monocytes with R3 neutralizes C1q-mediated enhancement of phagocytosis. In addition, when immobilized, R3 mimics C1q- MBL- or SPA-mediated enhancement of phagocytosis as reported. CDw93 (C1qRp) has been reported to define a human stem cell population with hematopoietic and hepatic potential.
Host Species: Mouse
Clone: R3
Isotype: IgM, κ
Species Reactivity: Human
Immunogen: C1q-binding protein
Flow Cytometry, Immunoprecipitation, Western Blotting

Specifications
CDw93 (C1qRp) | |
Monoclonal | |
0.5mg/mL | |
C1qRp | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
R3 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
C1q-binding protein | |
0.1 mg | |
Cell Biology | |
Human | |
IgM κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.