Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEA cell adhesion molecule 16 (Ceacam16) Rabbit anti-Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABN190
Description
This Anti-CEA cell adhesion molecule 16 (Ceacam16) Antibody is validated for use in Western Blotting, immunohistochemistry, Immunofluorescence for the detection of CEA cell adhesion molecule 16 (Ceacam16).
Specifications
CEA cell adhesion molecule 16 (Ceacam16) | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Mouse |
Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
Ceacam16 | |
Linear peptide corresponding to mouse Ceacam16 monomers and dimers. | |
100 μg | |
NM_001033419 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction