Learn More
CCAAT/enhancer binding protein (C/EBP), gamma, Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
The C/EBP family of transcription factors regulates viral and cellular CCAAT/enhancer element-mediated transcription. C/EBP proteins contain the bZIP region, which is characterized by two motifs in the C-terminal half of the protein: a basic region involved in DNA binding and a leucine zipper motif involved in dimerization. The C/EBP family consist of several related proteins, C/EBP alpha, C/EBP beta, C/EBP gamma, and C/EBP delta, that form homodimers and that form heterodimers with each other. CCAAT/enhancer binding protein gamma may cooperate with Fos to bind PRE-I enhancer elements. [provided by RefSeq
Specifications
Specifications
| Antigen | CCAAT/enhancer binding protein (C/EBP), gamma |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a full-length recombinant CEBPG. |
| Formulation | 50% glycerol |
| Gene | CEBPG |
| Gene Accession No. | BC013128 |
| Gene Alias | GPE1BP/IG/EBP-1 |
| Gene Symbols | CEBPG |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.