Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Surface Antigen of Human Nucleated Erythroid Cells Mouse anti-Human, Clone: HAE9, MilliporeSigma™

Catalog No. MAB2115MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2115MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2115MI Supplier MilliporeSigma Supplier No. MAB2115
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Cell Surface Antigen of Human Nucleated Erythroid Cells Antibody, clone HAE9 is validated for use in flow cytometry, immunoprecipitation for the detection of Cell Surface Antigen of Human Nucleated Erythroid Cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cell Surface Antigen of Human Nucleated Erythroid Cells
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone HAE9
Conjugate Unconjugated
Formulation Purified immunoglobulin via gel filtration from tissue culture supernatants in 20mM PB pH 7.6, 250mM NaCL, with 0.1% sodium azide as a preservative.
Host Species Mouse
Immunogen Human fetal liver cells
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for up to 6 months.
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.