Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CELSR1 Rabbit anti-Rat, Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABT119
Description
This Anti-CELSR1 Antibody is validated for use in Western Blotting, immunohistochemistry, Immunofluorescence for the detection of CELSR1.
Specifications
CELSR1 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat |
Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
CELSR1; CDHF9; FMI2 | |
Linear peptide corresponding to human CELSR1. | |
100 μg | |
NP_055061 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction