Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CELSR3, Mouse, Clone: 2F7, Abnova™

Catalog No. 89021401 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-401 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-401 Supplier Abnova Corporation Supplier No. H00001951M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CELSR3.

The protein encoded by this gene is a member of the flamingo subfamily, part of the cadherin superfamily. The flamingo subfamily consists of nonclassic-type cadherins; a subpopulation that does not interact with catenins. The flamingo cadherins are located at the plasma membrane and have nine cadherin domains, seven epidermal growth factor-like repeats and two laminin A G-type repeats in their ectodomain. They also have seven transmembrane domains, a characteristic unique to this subfamily. It is postulated that these proteins are receptors involved in contact-mediated communication, with cadherin domains acting as homophilic binding regions and the EGF-like domains involved in cell adhesion and receptor-ligand interactions. The specific function of this particular member has not been determined. [provided by RefSeq

Sequence: REDGGPGLGVREPIFVGLRGRRQSARNSRGPPEQPNEELGIEHGVQPLGSRERETGQGPGSVLYWRPEVSSCGRTGPLQRGSLSPGALSSGVPGSGNSSPLPSDFLIRHH

Specifications

Antigen CELSR3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2F7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CELSR3.
Formulation PBS with no preservative; pH 7.4
Gene CELSR3
Gene Accession No. NM_001407
Gene Alias CDHF11/EGFL1/FMI1/HFMI1/MEGF2/RESDA1
Gene Symbols CELSR3
Host Species Mouse
Immunogen CELSR3 (NP_001398, 71 a.a. ∼ 180 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1951
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.