Learn More
centromere protein J, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human CENPJ protein.
Supplier: Abnova Corporation H00055835D01P
Description
This gene encodes a protein that belongs to the centromere protein family. During cell division, this protein plays a structural role in the maintenance of centrosome integrity and normal spindle morphology, and it is involved in microtubule disassembly at the centrosome. This protein can function as a transcriptional coactivator in the Stat5 signaling pathway, and also as a coactivator of NF-kappaB-mediated transcription, likely via its interaction with the coactivator p300/CREB-binding protein. Mutations in this gene are associated with primary autosomal recessive microcephaly, a disorder characterized by severely reduced brain size and mental retardation. [provided by RefSeq
Sequence: MRKLQKERKVFEKYTTAARTFPDKKEREEIQTLKQQIADLREDLKRKETKWSSTHSRLRSQIQMLVRENTDLREEIKVMERFRLDAWKRAEAIESSLEVEKKDKLANTSVRFQNSQISSGTQVEKYKKNYLPMQGNPPRRSKSAPPRDLGNLDKGQAASPREPLEPLNFPDPEYKEEEEDQDIQGEISHPDGKVEKVYKNGCRVILFPNGTRKEVSADGKTITVTFFNGDVKQVMPDQRVIYYYAAAQTTHTTYPEGLEVLHFSSGQIEKHYPDGRKEITFPDQTVKNLFPDGQEESIFPDGTIVRVQRDGNKLIEFNNGQRELHTAQFKRREYPDGTVKTVYANGHQETKYRSGRIRVKDKEGNVLMDTELSpecifications
centromere protein J | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human CENPJ protein. | |
CENPJ | |
BM032/CPAP/LAP/LIP1/MCPH6/MGC131581/MGC131582/MGC142222/MGC142224 | |
Rabbit | |
Affinity Purified | |
RUO | |
55835 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC024209.2 | |
CENPJ | |
CENPJ (AAH24209.3, 1 a.a. ∼ 372 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.