Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CENPO Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520826
Description
A suggested positive control is mouse kidney tissue lysate. PA5-20826 can be used with blocking peptide PEP-0940.
Accurate chromosome segregation during mitosis requires a kinetochore to assemble correctly at the centromere of each chromatid and form a dynamic interface with the microtubules of the mitotic spindle. The kinetochore assembly includes the multisubunit CENP-H/I complex which can be divided into three functional classes based on phenotype analysis. One of these classes is referred to as the CENP-O class of proteins of which CENPO is the exemplar. These proteins form a stable complex with each other and are required for proper kinetochore function and for recovery from mitotic spindle damage. CENPO has also been shown to be an auto-antigen in a small population of anti-centromere antibodies- (ACA-) positive patients with sclerodoma.
Specifications
CENPO | |
Polyclonal | |
Unconjugated | |
CENPO | |
2810429O05Rik; 8430427C03Rik; CENPO; CENP-O; Centromere protein O; centromeric protein O; D12Ertd482e; ICEN36; ICEN-36; Interphase centromere complex protein 36; MCM21R | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
52504, 684439, 79172 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8K015, Q9BU64 | |
CENPO | |
A 16 amino acid peptide from near the center of human CENPO. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction