Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

centromere protein P, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018707 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-707 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-707 Supplier Abnova Corporation Supplier No. H00401541D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human CENPP protein.

CENPP is a subunit of a CENPH (MIM 605607)-CENPI (MIM 300065)-associated centromeric complex that targets CENPA (MIM 117139) to centromeres and is required for proper kinetochore function and mitotic progression (Okada et al., 2006 [PubMed 16622420]).[supplied by OMIM

Sequence: MDAELAEVRALQAEIAALRRACEDPPAPWEEKSRVQKSFQAIHQFNLEGWKSSKDLKNQLGHLESELSFLSTLTGINIRNHSKQTEDLTSTEMTEKSIRKVLQRHRLSGNCHMVTFQLEFQILEIQNKERLSSAVTDLNIIMEPTECSELSEFVSRAEERKDLFMFFRSLHFFVEWFEYRKRTFKHLKEKYPDAVYLSEGPSSCSMGIRSASRPGFELVIVWRIQIDEDGKVFPKLDLLTKVPQRALELDKNRAIETAPLSFRTLVGLLGIEAALESLIKSLCAEENN

Specifications

Antigen centromere protein P
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human CENPP protein.
Formulation PBS with no preservative; pH 7.4
Gene CENPP
Gene Accession No. NM_001012267.1
Gene Alias CENP-P/FLJ33928
Gene Symbols CENPP
Host Species Rabbit
Immunogen CENPP (NP_001012267.1, 1 a.a. ∼ 288 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401541
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.