Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ArfGAP with dual PH domains 2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89007897 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-007-897 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-007-897 Supplier Abnova Corporation Supplier No. H00055803B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human CENTA2 protein.

Sequence: MGDRERNKKRLLELLRAPDTGNAHCADCGAADPDWASYKLGIFICLNCCGVHRNFPDISRVKSVRLDFWDDSIVEFMIHNGNLRVKAKFEARVPAFYYIPQANDCLVLKEQWIRAKYERREFMADGETISLPGNREGFLWKRGRDNSQFLRRKFVLLAREGLLKYFTKEQGKSPKAVISIKDLNATFQTEKIGHPHGLQITYRRDGHTRNLFVYHESGKEIVDWFNALRAARLQYLKMAFPELPESELVPFLTRNYLKQGFMEKTGPKKEPFKKRWFALDCHERRLLYYKNPLDAFEQGQVFLGNKEQGYEAYEDLPKGIRGNRWKAGLTIVTPERRFVLTCPSEKEQQEWLESLRGVLSSPLTPLNRLTASTESGRSSR

Specifications

Antigen ArfGAP with dual PH domains 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human CENTA2 protein.
Formulation No additive
Gene ADAP2
Gene Accession No. BC033758
Gene Alias CENTA2/HSA272195/cent-b
Gene Symbols ADAP2
Host Species Mouse
Immunogen CENTA2 (AAH33758, 1 a.a. ∼ 380 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 55803
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.