Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Centrin Mouse anti-Human, Mouse, Rat, Clone: 20H5, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 041624
Description
Anti-Centrin Antibody, clone 20H5 is a Mouse Monoclonal Antibody for detection of Centrin also known as Caltractin isoform 2, EF-hand protein and has been validated in western blot, immunoprecipitation, immunocytochemistry.
Specifications
Centrin | |
Monoclonal | |
Unconjugated | |
NP_004057 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG2a κ |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
20H5 | |
Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
CEN1, CETN, Centrin-1 | |
Recombinant protein corresponding to the C-terminus of Chlamydomonas Centrin. | |
100 μg | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction