Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-CEP Clone: 7E12H12,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABC530
Description
Specifically detects CEP Clone: 7E12H12 in Human samples, and it is validated for Dot Blot, ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting
The Monoclonal (mAb) Das-1 was originally desiganted as clone 7E12H12 and was thought to target a 40kDa colon-specific tropomyosin (TM) isoform or a TM-related protein. It was later found that this clone specifically immunoprecipitated a high molecular weight chaperone protein that targets tropomyosin 5 (TM5) in human colon epithelial cells and subsequently named Colon Epithelial Protein (CEP). The Das-1 target is a glycoprotein with variable apparent sizes depending on the level of glycosylation. The target glycoprotein immunoprecipitated from the plasma membrane of DLD-1 colon cancer cells appears with an apparent molecular size of 185kDa, while the target isolated from LS-180 colon cancer cells migrates with a size range extends to greater than 200kDa. Das-1/CEP immunoreactivity is reported to be associated with Barrett's metaplasia, Barrett's adenocarcinoma, as well as with gastric adenocarcinomas and carcinoma-associated intestinal metaplasia, but no reactivity has been found in normal esophageal, gastric or small intestinal mucosa. When tested by ELISA and imunoblotting against recombinant human tropomyosins isoforms 1-5, none of five hTM isoforms reacted with mAB DAS-1. As determined by immunofluorescence and FACS analyses, CEP/Das-1 immunoreactivity is detected in several colon cancer lines (DLD-1, LS180, T84), but not in the colorectal adenocarcinoma cell line HT-29. CEP/Das-1 immunoreactivity is not detected among non-colonic epithelial cell lines (293-T, HeLa, pancreatic cancer) or hematopoietic lines (K562, KGI, Daudi).
Specifications
CEP | |
Monoclonal | |
Unconjugated | |
Mouse | |
Purified by ion-exchange chromatography | |
RUO | |
Primary | |
Human | |
Purified |
Dot Blot, ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
7E12H12 | |
Purified mouse monoclonal IgMκ antibody in PBS with 0.05% sodium azide. | |
Human colonic protein extract. | |
100 μL | |
Apoptosis, Cancer | |
Clone 7E12H12 (mAb DAS-1) specifically reacts with Colon Epithelial Protein (CEP), but not human tropomyosins isoforms (hTM1-5). CEP/Das-1 immunoreactivity is detected in several colon cancer lines (DLD-1, LS180, T84), but not in the colorectal adenocarcinoma cell line HT-29. CEP/Das-1 immunoreactivity is not detected among non-colonic epithelial cell lines (293-T, HeLa, HUVEC, pancreatic cancer) or hematopoietic lines (K562, KGI, Daudi). | |
2°C to 8°C for one year from date of shipment | |
IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction