Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceruloplasmin Mouse, Unlabeled, Clone: 8, BD

Catalog No. BDB611489 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611489 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611489 Supplier BD Biosciences Supplier No. 611489

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Ceruloplasmin (CP) is a copper-containing glycoprotein that carries approximately 90% of plasma copper. CP contains six copper atom binding sites and is prone to transfer its copper atoms to tissues, however it is not essential for copper transport. In cultured endothelial cells, CP release of copper in the cytosol inhibits nitric oxide synthase activation, and this may be important for regulating vascular tone in blood vessels. In addition to its copper binding role, CP has also been shown to have ferroxidase activity, which involves conversion of ferrous to
ferric iron. The iron metabolism disorder aceruloplasminemia results from mutations in CP and is characterized by massive iron deposits in liver, pancreas, brain, retina, and other tissues. CP is expressed in the liver, the splenic reticuloendothelial system, the bronchiolar epithelium of the lung, and the retina and brain. The cell-specific expression of CP in neural tissues may account for the neuropathologies associated with aceruloplaminemia, while the wide expression pattern of CP may demonstrate its importance in the regulation of copper and iron levels in many tissues.

Host Species: Mouse
Clone: 8
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Mouse ceruloplasmin aa. 233-355
Formula Weight [Chemical]: 132kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ceruloplasmin
Applications Western Blot
Classification Monoclonal
Clone 8
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse ceruloplasmin aa. 233-355
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.