Learn More
Anti- Cetuximab (Anti-Idiotype) Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody
Supplier:  R&D Systems FAB9626V100UG
Catalog No. FAB9626V100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Cetuximab Monoclonal specifically detects Cetuximab in Multi-Species samples. It is validated for Flow Cytometry.
Specifications
Specifications
Cetuximab
Monoclonal
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Full length Cetuximab,
100 μg
Primary
Multi-species
Purified
Flow Cytometry
2259C
Flow Cytometry 0.25-1 ug/10^6 cells
Rabbit
Protein A or G purified from cell culture supernatant
RUO
Detects Cetuximab in Western blots and direct ELISAs. No cross-reactivity with Adalimumab, Rituximab, Trastuzumab, Brentuximab, Gemtuzumab, Ibritumomab, Tositumomab, Solanezumab, Teprotumumab, and Aducanumab is observed.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.