Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CFTR Mouse anti-Human, Clone: M3A7, MilliporeSigma™

Catalog No. 05583MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-583-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-583-MI Supplier MilliporeSigma Supplier No. 05583
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect CFTR using this Anti-CFTR Antibody, clone M3A7 validated for use in immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CFTR
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone M3A7
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in buffer containing 0.1MTris-glycine, pH 7.4, 0.15MNaCl, 0.05% sodium azide and 30% glycerol.
Gene Accession No. NM_000492
Gene Symbols CFTR, CFTR/MRP, ABCC7, CBAVD, dJ760C5.1, CF, MRP7, ABC35, TNR-CFTR
Host Species Mouse
Immunogen GST-fusion protein corresponding to residues 1197-1480 of human cystic fibrosis transmembrane conductance regulator (CFTR). The epitope maps within amino acids 1365-1395. Clone M3A7.
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.