Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CFTR Mouse anti-Human, Clone: MM13-4, MilliporeSigma™

Catalog No. 05581MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-581-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-581-MI Supplier MilliporeSigma Supplier No. 05581
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CFTR Antibody, clone MM13-4 detects level of CFTR and has been published and validated for use in immunocytochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CFTR
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MM13-4
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1k in buffer containing 0.02M phosphate buffer, pH 7.6, 0.25M NaCl, and 0.1% sodium azide.
Gene Accession No. NM_000492
Gene Symbols CFTR, CFTR/MRP, ABCC7, CBAVD, dJ760C5.1, CF, MRP7, ABC35, TNR-CFTR
Host Species Mouse
Immunogen Synthetic peptide (RKGYRQRLELSD) corresponding to residues 25-36 of human cystic fibrosis transmembrane conductance regulator (CFTR). The immunizing sequence is identical in monkey and rabbit, has 11/12 identical amino acids in sheep, cow and frog, 10/12
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.