Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hCG beta, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA127846
Description
This protein is a member of the glycoprotein hormone beta chain family and encodes the beta 3 subunit of chorionic gonadotropin (CG). Glycoprotein hormones are heterodimers consisting of a common alpha subunit and an unique beta subunit which confers biol
PA1-27846 detects CGB from human samples.PA1-27846 may be successfully used in Western blot applications.The PA1-27870 immunogen is a synthetic peptide corresponding to human Chorionic gonadotropin beta subunit.
Specifications
CGB | |
Polyclonal | |
CGB3 | |
CGB3, hCGB | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
1082 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Western Blot | |
Unconjugated | |
P0DN86 | |
CGB3 | |
Synthetic peptide corresponding to human Chorionic gonadotropin beta subunit. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction