Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cGMP (cyclic GMP) Rabbit anti-All, Polyclonal, MilliporeSigma™

Catalog No. 09101MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-101-MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-101-MI Supplier MilliporeSigma Supplier No. 09101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-cGMP (cyclic GMP) Antibody detects level of cGMP (cyclic GMP) and has been published and validated for use in ELISA, immunohistochemistry, RIA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen cGMP (cyclic GMP)
Applications ELISA, Immunohistochemistry, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Gene Symbols PRKG1; cGKI-beta; FLJ36117; PRKG1B; PGK; cGKI-BETA; PRKGR1A; cGKI-alpha; PRKGR1B; cGKI; MGC71944; DKFZp686K042; OTTHUMP00000061216; CGKI
Host Species Rabbit
Immunogen 2 Monosuccinyl cyclic GMP conjugated to KLH
Purification Method Serum
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001098512.1; NM_006258.2
Target Species All species
Content And Storage −20°C for 24 months
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.