Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CGRP Monoclonal Antibody (4901)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA120209
Description
Recommended positive controls: rat alpha-CGRP. MA1-20209 has been successfully used in ELISA, radioimmune assay, neutralizing, and immunocytochemistry procedures. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
CGRP | |
Monoclonal | |
2.1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P01256, P01257, P01258, P06881, P41547 | |
Calca | |
Rat alpha-CGRP peptide. | |
50 μL | |
Primary | |
Human, Rat, Canine | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry, Neutralization, Radioimmune Assays (RIA), Immunocytochemistry | |
4901 | |
Unconjugated | |
Calca | |
alpha-type CGRP; CA; Cal1; CAL6; CALC; Calc1; Calca; calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; Calcitonin gene-related peptide 1; calcitonin gene-related peptide I; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-related polypeptide alpha; CCALCI; CCP; Cgrp; CGRP1; CGRP-1; CGRP-I; Ct; Ctn; Katacalcin; KC; MGC126648; PCT; PDN-21; prepro-alpha-calcitonin gene-related peptide; RAT calcitonin; RATCAL6 | |
Mouse | |
purified | |
RUO | |
24241, 403946, 796 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction