Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHAD, Mouse, Clone: 8B7, Abnova™

Catalog No. 89021065 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-065 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-065 Supplier Abnova Corporation Supplier No. H00001101M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CHAD.

Chondroadherin is a cartilage matrix protein thought to mediate adhesion of isolated chondrocytes. The protein contains 11 leucine-rich repeats flanked by cysteine-rich regions. The chondroadherin messenger RNA is present in chondrocytes at all ages. [provided by RefSeq

Sequence: LDNTNLEKFSDGAFLGVTTLKHVHLENNRLNQLPSNFPFDSLETLALTNNPWKCTCQLRGLRRWLEAKASRPDATCASPAKFKGQHIRDTDAFRSCKFPTKRSKKAGRH

Specifications

Antigen CHAD
Applications ELISA
Classification Monoclonal
Clone 8B7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CHAD.
Formulation PBS with no preservative; pH 7.4
Gene CHAD
Gene Accession No. NM_001267
Gene Alias SLRR4A
Gene Symbols CHAD
Host Species Mouse
Immunogen CHAD (NP_001258, 251 a.a. ∼ 359 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1101
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.