Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD3 Mouse, Unlabeled, Clone: 44, BD

Catalog No. BDB611846 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611846 50 μg
BDB611847 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611846 Supplier BD Biosciences Supplier No. 611846

Mouse Monoclonal Antibody

The SNF2-related family of proteins all contain conserved helicase motifs, and have been implicated in transcription regulation, DNA repair, and DNA recombination. The DNA-dependent ATPase activity of these proteins is usually activated by single-stranded, double-stranded, or nucleosomal DNA. The chromodomain helicase DNA binding proteins (CHDs) are a subfamily of the SNF2 family, and includes CHD1, CHD2, CHD3, and CHD4. CHD3 and CHD4 contain N-terminal paired PHD zinc finger motifs (PZF), two heterochromatin binding domains (chromodomains; CDs), and a central SNF2-like helicase/ATPase domain. CHD3 mRNA is widely expressed, and CHD3 protein localizes to the nucleus, but not nucleoli. Both CHD3 and CHD4 were first identified as self-antigens recognized by sera from patients with the autoimmune connective-tissue disease Mi-2 dermatomyositis. In addition, both CHD3 and CHD4 are associated with a nucleosome remodelling and deacetylating (NRD) complex, and facilitate deacetylation of oligonucleosomal histones in vitro. Thus, CHD3 and CHD4 are chromodomain containing proteins that may have important roles in ATP-dependent nucleosome remodelling.

Host Species: Mouse
Clone: 44
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human CHD3 aa. 1277-1474

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD3
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 44
Concentration 250μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human CHD3 aa. 1277-1474
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.